Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114018
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114018 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114018 Supplier Invitrogen™ Supplier No. PA5114018
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKTTSDRSRE QTNTRSKHME KLEVDPSKHS FPDVTNIVQE KKHTPRRRPE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 79%; Dog: 79%; Horse: 86%; Human: 100%; Mouse: 92%; Pig: 93%; Rabbit: 100%; Rat: 100%.

LRRC6 may be involved in spermatocytogenesis or prophase of meiosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LRRC6
Gene Accession No. Q86X45
Gene Alias CILD19; leucine rich repeat containing 6; leucine rich repeat containing 6 (testis); leucine-rich repeat-containing 6 (testis); leucine-rich repeat-containing protein 6; leucine-rich repeat-containing protein 6; protein tilB homolog; leucine-rich testis-specific protein; LRRC6; Lrtp; Mc2; Protein tilB homolog; testis-specific leucine-rich repeat protein; Tslrp; Unknown (protein for MGC:138048)
Gene Symbols LRRC6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human LRRC6.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23639
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.