Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC61 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116611
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116611 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116611 Supplier Invitrogen™ Supplier No. PA5116611
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse eye, Mouse brain, Rat brain Immunogen sequence: MDPPAEKPGE AGGLQITPQL LKSRTGEFSL ESILLLKLRG LGLADLGCLG ECLGLEWLDL SGNALTHLGP LASLRQLAVL NVSNNRLTGL EPLATCENLQ SLNAAGNLLA TPGQLQCLAG LPCLEYLRLR DPLARLSNPL CANPSYWAAV RELLPGLKVI DGERVIGRGS EFYQLCRDLD SSLRPSSSPG PRATEAQPWV EPGYWESWPS RSSSILEEAC RQFQDTLQEC WDLDRQASDS LAQAEQVLSS AGPTSSFVF.

TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC61
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LRRC61
Gene Accession No. Q8R2R5
Gene Alias BC027309; HSPC295; leucine rich repeat containing 61; leucine-rich repeat-containing protein 61; Lrrc61; RGD1561800
Gene Symbols LRRC61
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-259 of human LRRC61 (NP_0764311).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 243371, 500111
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.