Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC66 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555710
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555710 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555710 Supplier Invitrogen™ Supplier No. PA555710
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QRNKLSDTPK GLWKLKSLQS LDLSFNGILQ IGWSDFHNCL QLENLCLKSN KIFKIPPQAF KDLKKLQVID LSNNALITIL PMMIIALEFP HLVVDLADNN WQCDDSVAVF QNFISESWRK KWNVICNRSI Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 58%.

This gene encodes a protein belonging to the leucine-rich repeat family of proteins, which are involved in diverse biological processes, including cell adhesion, cellular trafficking, and hormone-receptor interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC66
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRRC66
Gene Accession No. Q68CR7
Gene Alias leucine rich repeat containing 66; leucine-rich repeat-containing protein 66; LRRC66
Gene Symbols LRRC66
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRC66. Recombinant protein control fragment (Product #RP-91061).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339977
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.