Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC67 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS18400R

Description
Regulates phosphatase activity of protein phosphatase 1 (PP1) complexes in the testis.Specifications
LRRC67 | |
Polyclonal | |
Unconjugated | |
PPP1R42 | |
dtr; leucine rich repeat containing 67; Leucine-rich repeat-containing protein 67; LRRC67; PPP1R42; Protein phosphatase 1 regulatory subunit 42; protein phosphatase 1, regulatory subunit 42; testis leucine-rich repeat; TLLR | |
Rabbit | |
Protein A | |
RUO | |
286187 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q7Z4L9 | |
PPP1R42 | |
KLH conjugated synthetic peptide derived from rat Nestin. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction