Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRC69 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561217
Description
Immunogen sequence: LIKALSGGKN TKIITLNGKK MTKMPSALGK LPGLKTLVLQ NNLIPKVCPE LCNLTQFQDL KLREFYCEGN PLFLQQPVIS TQQENVWSLQ EIT Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 51%.

Specifications
LRRC69 | |
Polyclonal | |
Unconjugated | |
LRRC69 | |
leucine rich repeat containing 69; leucine-rich repeat-containing protein 69; LRRC69 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100130742 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZNQ3 | |
LRRC69 | |
Recombinant protein corresponding to Human LRRC69. Recombinant protein control fragment (Product #RP-100338). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction