Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC69 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549157
Description

Specifications
LRRC69 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q6ZNQ3 | |
LRRC69 | |
KLH conjugated synthetic peptide between 200-227 amino acids from the Central region of human LRRC69. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
LRRC69 | |
Leucine-rich repeat-containing protein 69, LRRC69 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100130742 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction