Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570660
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570660 Supplier Invitrogen™ Supplier No. PA570660
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 93%; Rat: 93%.

LRRC7 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LRRC7
Gene Accession No. Q96NW7
Gene Alias AK122498; densin; densin 13T-16; densin 13T-17; densin 13T-23; densin 13T-29; densin 13T-41; densin 13T-43; densin 13T-57; densin 13T-57C; densin 13T-72; densin 13T-89; densin 180; densin-180; Kiaa1365; Lap1; leucine rich repeat containing 7; leucine-rich repeat-containing protein 7; LOW QUALITY PROTEIN: leucine-rich repeat-containing protein 7; Lrrc7; mKIAA1365; Protein LAP1
Gene Symbols LRRC7
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human LRRC7 (aa 1235-1284).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57554
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.