Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC70 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553056
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553056 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553056 Supplier Invitrogen™ Supplier No. PA553056
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLSSLIHLQA NSNPWECNCK LLGLRDWLAS SAITLNIYCQ NPPSMRGRAL RYINITNCVT SSINVSRAWA VVKSPHIHHK TTALMMAWHK VTTNGSPLEN TETENITFWE RIPTSPAGRF FQENAFGNPL ETTAVLPVQI QL Highest antigen sequence identity to the following orthologs: Mouse - 21%, Rat - 19%.

Renders cells highly sensitive to the activation by cytokines and lipopolysaccharide (LPS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC70
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRRC70
Gene Accession No. Q7Z2Q7
Gene Alias leucine rich repeat containing 70; leucine-rich repeat-containing protein 70; LRRC70; SLRN; Synleurin; UNQ1891/PRO4337
Gene Symbols LRRC70
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRC70. Recombinant protein control fragment (Product #RP-90563).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100130733
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.