Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC75A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571039
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571039 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571039 Supplier Invitrogen™ Supplier No. PA571039
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRRC75A Polyclonal Antibody for Western Blot

LRRC75A is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC75A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LRRC75A
Gene Accession No. Q8NAA5
Gene Alias C17orf76; FAM211A; family with sequence similarity 211, member A; leucine rich repeat containing 75A; leucine-rich repeat-containing protein 75A; leucine-rich repeat-containing protein C17orf76; leucine-rich repeat-containing protein FAM211A; LRRC75A
Gene Symbols LRRC75A
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human LRRC75A.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388341
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.