Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRC8A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572870
Description
LRRC8A Polyclonal Antibody for Western Blot, IHC (P), ELISA
Essential component of the volume-regulated anion channel (VRAC, also named VSOAC channel), an anion channel required to maintain a constant cell volume in response to extracellular or intracellular osmotic changes. The VRAC channel conducts iodide better than chloride and may also conduct organic osmolytes like taurine. It is unclear whether LRRC8A constitutes a pore-forming subunit or whether it is closely associated with the pore and mediates channel properties such as ion selectivity. Involved in B-cell development: required for the pro-B cell to pre-B cell transition. Also required for T-cell development.
Specifications
LRRC8A | |
Polyclonal | |
Unconjugated | |
LRRC8A | |
AGM5; KIAA1437; leucine rich repeat containing 8 family member A; leucine rich repeat containing 8 family, member A; leucine rich repeat containing 8A; leucine-rich repeat containing 8 family member A; leucine-rich repeat-containing 8; leucine-rich repeat-containing protein 8A; Lrrc8; LRRC8A; mKIAA1437; SWELL1; Swelling protein 1; UNQ221/PRO247; Volume-regulated anion channel subunit LRRC8A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
241296, 311846, 56262 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q4V8I7, Q80WG5, Q8IWT6 | |
LRRC8A | |
LRRC8A antibody for a 15 amino acid peptide near the amino terminus of human LRRC8A. The immunogen is within amino acids 60 - 110 of LRRC8A. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction