Learn More
LRRC8A Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304175AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat
Essential component of the volume-regulated anion channel (VRAC, also named VSOAC channel), an anion channel required to maintain a constant cell volume in response to extracellular or intracellular osmotic changes. The VRAC channel conducts iodide better than chloride and may also conduct organic osmolytes like taurine. It is unclear whether LRRC8A constitutes a pore-forming subunit or whether it is closely associated with the pore and mediates channel properties such as ion selectivity. Involved in B-cell development: required for the pro-B cell to pre-B cell transition. Also required for T-cell development.Specifications
LRRC8A | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q80WG5, Q8IWT6 | |
Lrrc8a | |
Between 760 and 810 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
LRRC8A | |
Volume-regulated anion channel subunit LRRC8A, Leucine-rich repeat-containing protein 8A, Swelling protein 1, LRRC8A, KIAA1437, LRRC8, SWELL1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
241296, 56262 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.