Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC8A Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501567050
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7050 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7050 Supplier Bethyl Laboratories, Inc Supplier No. A304175A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat

Essential component of the volume-regulated anion channel (VRAC, also named VSOAC channel), an anion channel required to maintain a constant cell volume in response to extracellular or intracellular osmotic changes. The VRAC channel conducts iodide better than chloride and may also conduct organic osmolytes like taurine. It is unclear whether LRRC8A constitutes a pore-forming subunit or whether it is closely associated with the pore and mediates channel properties such as ion selectivity. Involved in B-cell development: required for the pro-B cell to pre-B cell transition. Also required for T-cell development.

Specifications

Antigen LRRC8A
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene LRRC8A
Gene Accession No. Q80WG5, Q8IWT6
Gene Alias Volume-regulated anion channel subunit LRRC8A, Leucine-rich repeat-containing protein 8A, Swelling protein 1, LRRC8A, KIAA1437, LRRC8, SWELL1
Gene Symbols LRRC8A
Host Species Rabbit
Immunogen Between 760 and 810
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 241296, 56262
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.