Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC8B Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297498AF350
Description
LRRC8B Polyclonal specifically detects LRRC8B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
LRRC8B | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
KIAA0231TA-LRRPleucine-rich repeat-containing protein 8B, leucine rich repeat containing 8 family, member B, MGC42220, T cell activation leucine repeat rich protein, TALRRP, T-cell activation leucine repeat-rich protein | |
Produced in rabbits immunized with E. coli-derived Human LRRC8B fragment. | |
0.1 mL | |
Stem Cell Markers | |
23507 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction