Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC8B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | LRRC8B |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Description
LRRC8B Polyclonal specifically detects LRRC8B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LRRC8B | |
Unconjugated | |
RUO | |
KIAA0231TA-LRRPleucine-rich repeat-containing protein 8B, leucine rich repeat containing 8 family, member B, MGC42220, T cell activation leucine repeat rich protein, TALRRP, T-cell activation leucine repeat-rich protein | |
LRRC8B | |
IgG | |
92 kDa |
Polyclonal | |
Rabbit | |
Stem Cell Markers | |
23507 | |
Synthetic peptides corresponding to LRRC8B(leucine rich repeat containing 8 family, member B) The peptide sequence was selected from the middle region of LRRC8B. Peptide sequence TLYLKSSLSRIPQVVTDLLPSLQKLSLDNEGSKLVVLNNLKKMVNLKSLE. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title