Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC8B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$231.00 - $262.00
Specifications
Antigen | LRRC8B |
---|---|
Dilution | Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 uL/mg of lysate |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
LRRC8B Polyclonal antibody specifically detects LRRC8B in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
LRRC8B | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Stem Cell Markers | |
0.2 um filtered solution in PBS | |
23507 | |
IgG | |
Antigen and protein A Affinity-purified |
Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 uL/mg of lysate | |
Polyclonal | |
Purified | |
RUO | |
Human | |
KIAA0231TA-LRRPleucine-rich repeat-containing protein 8B, leucine rich repeat containing 8 family, member B, MGC42220, T cell activation leucine repeat rich protein, TALRRP, T-cell activation leucine repeat-rich protein | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human LRRC8B. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title