Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC8B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112101 Supplier Invitrogen™ Supplier No. PA5112101
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody reacts with: Human, Mouse (Species predicted to react based on 100% sequence homology).

Non-essential component of the volume-regulated anion channel (VRAC, also named VSOAC channel), an anion channel required to maintain a constant cell volume in response to extracellular or intracellular osmotic changes. The VRAC channel conducts iodide better than chloride and may also conduct organic osmolytes like taurine. Channel activity requires LRRC8A plus at least one other family member (LRRC8B, LRRC8C, LRRC8D or LRRC8E); channel characteristics depend on the precise subunit composition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC8B
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene LRRC8B
Gene Accession No. Q6P9F7
Gene Alias KIAA0231; leucine rich repeat containing 8 family member B; leucine rich repeat containing 8 family, member B; leucine-rich repeat containing 8 family member B; leucine-rich repeat-containing protein 8B; LRRC8B; T cell activation leucine repeat rich protein; TALRRP; TA-LRRP; T-cell activation leucine repeat-rich protein; UNQ6413/PRO21207; Volume-regulated anion channel subunit LRRC8B
Gene Symbols LRRC8B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human LRRC8B.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23507
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.