Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC8B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553675
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553675 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553675 Supplier Invitrogen™ Supplier No. PA553675
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSTSSRLEHF VAILHKCFDS PWTTRALSET VAEQSVRPLK LSKSKILLSS SGCSADIDSG KQSLPYPQPG LESAGIESPT SSVLDKKEGE QAKAIFEKVK RFRMHVEQKD Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

Non-essential component of the volume-regulated anion channel (VRAC, also named VSOAC channel), an anion channel required to maintain a constant cell volume in response to extracellular or intracellular osmotic changes. The VRAC channel conducts iodide better than chloride and may also conduct organic osmolytes like taurine. Channel activity requires LRRC8A plus at least one other family member (LRRC8B, LRRC8C, LRRC8D or LRRC8E); channel characteristics depend on the precise subunit composition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC8B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRRC8B
Gene Accession No. Q6P9F7
Gene Alias KIAA0231; leucine rich repeat containing 8 family member B; leucine rich repeat containing 8 family, member B; leucine-rich repeat containing 8 family member B; leucine-rich repeat-containing protein 8B; LRRC8B; T cell activation leucine repeat rich protein; TALRRP; TA-LRRP; T-cell activation leucine repeat-rich protein; UNQ6413/PRO21207; Volume-regulated anion channel subunit LRRC8B
Gene Symbols LRRC8B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRC8B. Recombinant protein control fragment (Product #RP-91195).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23507
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.