Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC8E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554218
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554218 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554218 Supplier Invitrogen™ Supplier No. PA554218
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLRQKLQRNA AGRLELALCM LPGLPDTVFE LSEVESLRLE AICDITFPPG LSQLVHLQEL SLLHSPARLP FSLQVFLRDH LKVMRVKCEE LREVPLWVFG LRGLEELHLE GLFPQELARA ATL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.

LRRC8 is a multi-pass membrane protein. It contains 13 LRR (leucine-rich) repeats. LRRC8 involved in B cell development belongs to a novel family of leucine-rich repeat proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC8E
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRRC8E
Gene Accession No. Q6NSJ5
Gene Alias 1810049O03Rik; C87354; FLJ23420; leucine rich repeat containing 8 family member E; leucine rich repeat containing 8 family, member E; leucine rich repeat containing 8 VRAC subunit E; leucine-rich repeat containing 8 family member E; leucine-rich repeat-containing protein 8E; LOW QUALITY PROTEIN: volume-regulated anion channel subunit LRRC8E; Lrrc8e; RGD1311979; Volume-regulated anion channel subunit LRRC8E
Gene Symbols LRRC8E
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRC8E. Recombinant protein control fragment (Product #RP-91107).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80131
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.