Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583527
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583527 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583527 Supplier Invitrogen™ Supplier No. PA583527
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SFKELTNLTR LPCLKDLCLN DPQYTTNPVC LLCNYSTHVL YHLPCLQRFD TLDVSAKQIK ELADTTAMKK IMYYNMRIKT LQRHLKEDLE KLNDQKCKLQ KLPE Highest antigen sequence indentity to the following orthologs: Mouse - 86%, Rat - 86%.

LRRC9 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRRC9
Gene Accession No. Q6ZRR7
Gene Alias FLJ46156; leucine rich repeat containing 9; LRRC9
Gene Symbols LRRC9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRC9. Recombinant protein control fragment (Product #RP-98431).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 341883
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.