Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRCC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139937
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139937 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139937 Supplier Invitrogen™ Supplier No. PA5139937
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: KIPYDAKTIQ TIKHHNKNYN SFVSCNRKMK PPYLKELYVS SSLANCPMLQ ESEKPKTEII KVDQSHSEDN TYQSLVEQLD QEREKRWRAE QAENK Highest antigen sequence indentity to the following orthologs: Rat - 67%, Mouse - 65%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRCC1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRRCC1
Gene Accession No. Q9C099
Gene Alias centrosomal leucine-rich repeat and coiled-coil containing protein; centrosomal leucine-rich repeat and coiled-coil domain-containing protein; CLERC; KIAA1764; leucine rich repeat and coiled-coil centrosomal protein 1; leucine rich repeat and coiled-coil domain containing 1; leucine-rich repeat and coiled-coil domain-containing protein 1; LRRCC1; SAP2; sodium channel associated protein 2; variable number of flagella 1 homolog; VFL1
Gene Symbols LRRCC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRCC1. Recombinant protein control fragment (Product #RP-109510).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85444
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.