Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140355
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140355 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140355 Supplier Invitrogen™ Supplier No. PA5140355
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: NLTALPSAIY NIFSLKEINF DDNPLLRPPV EICKGKQLYT IARYLQRADE RDGRLK Highest antigen sequence indentity to the following orthologs: Mouse - 70%, Rat - 75%.

LRRD1 (Leucine Rich Repeats And Death Domain Containing 1) is a Protein Coding gene. An important paralog of this gene is SCRIB. Predicted to be involved in positive regulation of Ras protein signal transduction and signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRRD1
Gene Accession No. A4D1F6
Gene Alias Leucine-rich repeat domain-containing protein 1; Leucine-rich repeat protein 1
Gene Symbols LRRD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRD1. Recombinant protein control fragment (Product #RP-99977).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401387
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.