Learn More
LRRFIP1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303078AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Leucine-rich repeat FLI-I interacting protein 1 (LRRFIP1/FLAP-1) is a MyD88-interacting signal protein which mediates either induction or repression of TLR signaling. Flap-1 directly interacts with β-catenin and activates β-catenin-dependent transcription activity. Flap-1 or LRRFIP1 was also shown to bind the TNF- -308 promoter polymorphism site, suggesting its possible role in the regulation of TNF- production. FLAP-1 along with LRRFIP2 and FLIIH serve as a balance in coordinating inflammatory response after infection, and may act as a novel target in the control of agonist-induced TLR-mediated inflammatory and autoimmune diseases.Specifications
LRRFIP1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q32MZ4 | |
LRRFIP1 | |
Between 50 and 100 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
LRRFIP1 | |
FLAP-1, FLAP1, FLIIAP1, GCF-2, GCF2, HUFI-1, TRIP; GC-binding factor 2; LRR FLII-interacting protein 1; NEDD8-conjugating enzyme; TAR RNA-interacting protein; leucine-rich repeat flightless-interacting protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9208 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.