Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRFIP1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982243
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2243 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2243 Supplier Bioss Supplier No. BS12439R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Leucine-rich repeat FLI-I interacting protein 1 (LRRFIP1/FLAP-1) is a MyD88-interacting signal protein which mediates either induction or repression of TLR signaling. Flap-1 directly interacts with β-catenin and activates β-catenin-dependent transcription activity. Flap-1 or LRRFIP1 was also shown to bind the TNF- -308 promoter polymorphism site, suggesting its possible role in the regulation of TNF- production. FLAP-1 along with LRRFIP2 and FLIIH serve as a balance in coordinating inflammatory response after infection, and may act as a novel target in the control of agonist-induced TLR-mediated inflammatory and autoimmune diseases.

Specifications

Antigen LRRFIP1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LRRFIP1
Gene Accession No. Q32MZ4
Gene Alias AU024550; BOS_3897; Flap; FLAP (FLI LRR associated protein); FLAP1; Flap-1; FLIIAP1; FLI-LRR-associated protein 1; GC-binding factor 2; GCF2; GCF-2; H186 FLAP; HUFI1; HUFI-1; leucine rich repeat (in FLII) interacting protein 1; leucine rich repeat (in FLII) interacting protein 1-like; Leucine-rich repeat flightless-interacting protein 1; LOC100342928; LRR binding FLII interacting protein 1; LRR FLII-interacting protein 1; Lrrfip1; NEDD8-conjugating enzyme; TAR RNA-interacting protein; TRIP
Gene Symbols LRRFIP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LRRFIP1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9208
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.