Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRFIP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583587
Description
Immunogen sequence: VKTGEKHGLV IIPDGTPNGD VSHEPVAGAI TVVSQEAAQV LESAGEGPLD VRLRKLAGEK EELLSQIRKL KLQLEEERQK CSRNDGTVGD LA.
The leucine-rich repeat FLI-I-interacting protein 2 (LRRFIP2), like the related protein LRRFIP1, was identified in a yeast two-hybrid system through binding to the LRR domain of human FLI. It can activate the Wnt signaling pathway in cultured cells and is thought to be a component of the Wnt signaling pathway that modulates Wnt signaling through interactions with Disheveled to increase the cellular levels and transcription activity of beta-catenin. LRRFIP2 has recently been characterized as a positive regulator of the TLR4 signaling pathway for activating NF-kappa-B during the early host response to LPS stimulation through binding to the TLR adaptor protein MyD88, and that this interaction with MyD88 is governed by phosphorylation of specific residues in LRRFIP2.
Specifications
| LRRFIP2 | |
| Polyclonal | |
| Unconjugated | |
| LRRFIP2 | |
| 5133400F20Rik; AI850587; HUFI-2; leucine rich repeat (in FLII) interacting protein 2; leucine-rich repeat flightless-interactin protein 2-like protein; leucine-rich repeat flightless-interacting protein 2; LRR binding FLII interacting protein 2; LRR FLII-interacting protein 2; LRRFIP2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9209 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y608 | |
| LRRFIP2 | |
| Recombinant protein corresponding to Human LRRFIP2. Recombinant protein control fragment (Product #RP-104750). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction