Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRFIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599092
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599092 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599092 Supplier Invitrogen™ Supplier No. PA599092
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRRFIP2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The leucine-rich repeat FLI-I-interacting protein 2 (LRRFIP2), like the related protein LRRFIP1, was identified in a yeast two-hybrid system through binding to the LRR domain of human FLI. It can activate the Wnt signaling pathway in cultured cells and is thought to be a component of the Wnt signaling pathway that modulates Wnt signaling through interactions with Disheveled to increase the cellular levels and transcription activity of beta-catenin. LRRFIP2 has recently been characterized as a positive regulator of the TLR4 signaling pathway for activating NF-kappa-B during the early host response to LPS stimulation through binding to the TLR adaptor protein MyD88, and that this interaction with MyD88 is governed by phosphorylation of specific residues in LRRFIP2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRFIP2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LRRFIP2
Gene Accession No. Q9Y608
Gene Alias 5133400F20Rik; AI850587; HUFI-2; leucine rich repeat (in FLII) interacting protein 2; leucine-rich repeat flightless-interactin protein 2-like protein; leucine-rich repeat flightless-interacting protein 2; LRR binding FLII interacting protein 2; LRR FLII-interacting protein 2; LRRFIP2
Gene Symbols LRRFIP2
Host Species Rabbit
Immunogen Recombinant Human Leucine-rich repeat flightless-interacting protein 2 protein (211-342AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9209
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.