Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRFIP2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA521115

Catalog No. PIPA521115


Only null left
Add to Cart

Description

Description

A suggested positive control is rat colon lysate. PA5-21115 can be used with blocking peptide PEP-1229.

The leucine-rich repeat FLI-I-interacting protein 2 (LRRFIP2), like the related protein LRRFIP1, was identified in a yeast two-hybrid system through binding to the LRR domain of human FLI. It can activate the Wnt signaling pathway in cultured cells and is thought to be a component of the Wnt signaling pathway that modulates Wnt signaling through interactions with Disheveled to increase the cellular levels and transcription activity of beta-catenin. LRRFIP2 has recently been characterized as a positive regulator of the TLR4 signaling pathway for activating NF-kappa-B during the early host response to LPS stimulation through binding to the TLR adaptor protein MyD88, and that this interaction with MyD88 is governed by phosphorylation of specific residues in LRRFIP2.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LRRFIP2
Polyclonal
Unconjugated
LRRFIP2
5133400F20Rik; AI850587; HUFI-2; leucine rich repeat (in FLII) interacting protein 2; leucine-rich repeat flightless-interactin protein 2-like protein; leucine-rich repeat flightless-interacting protein 2; LRR binding FLII interacting protein 2; LRR FLII-interacting protein 2; LRRFIP2
Rabbit
Antigen Affinity Chromatography
RUO
301035, 71268, 9209
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q4V7E8, Q91WK0, Q9Y608
LRRFIP2
A 17 amino acid peptide near the carboxy terminus of human LRRFIP2.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.