Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRIQ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561169
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561169 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561169 Supplier Invitrogen™ Supplier No. PA561169
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATPDFVPEPS PHDLPMDEHV LPDDADINFG YCEVEEKCRQ SFEAWQEKQK ELEDKEKQTL KAQRDREEKQ FQEEEEKR Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 63%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRIQ1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRRIQ1
Gene Accession No. Q96JM4
Gene Alias KIAA1801; leucine rich repeats and IQ motif containing 1; leucine-rich repeat and IQ domain-containing protein 1; leucine-rich repeats and IQ motif containing 1; LRRIQ1; testicular tissue protein Li 111
Gene Symbols LRRIQ1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRIQ1. Recombinant protein control fragment (Product #RP-97733).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84125
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.