Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRIQ3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546528
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546528 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546528 Supplier Invitrogen™ Supplier No. PA546528
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LRTFSDIDKY YTEQKKQEYH KEKVRVVAMA QVARERVRVA VNEHLNQKKY.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRIQ3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LRRIQ3
Gene Accession No. A6PVS8
Gene Alias leucine rich repeat containing 44; leucine rich repeats and IQ motif containing 3; leucine-rich repeat and IQ domain-containing protein 3; Leucine-rich repeat-containing protein 44; leucine-rich repeats and IQ motif containing 3; LRRC44; Lrriq3
Gene Symbols LRRIQ3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human LRRIQ3 (aa 417-466).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127255
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.