Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRIQ4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139792
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139792 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139792 Supplier Invitrogen™ Supplier No. PA5139792
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: TDLKEIPVVI FKNLHHLELL GLTGNHLKCL PKEIVNQTKL REIYLKRNQF EVFPQELCVL YTLEIIDLDE NKIGAIPEEI GHLTGLQKFY MASNNL Highest antigen sequence indentity to the following orthologs: Rat - 70%, Mouse - 71%.

LRRIQ4 (Leucine Rich Repeats And IQ Motif Containing 4) is a Protein Coding gene. Diseases associated with LRRIQ4 include Retinitis Pigmentosa 38. An important paralog of this gene is LRRK1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRIQ4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRRIQ4
Gene Accession No. A6NIV6
Gene Alias leucine rich repeat containing 64; leucine rich repeats and IQ motif containing 4; leucine-rich repeat and IQ domain-containing protein 4; Leucine-rich repeat-containing protein 64; leucine-rich repeats and IQ motif containing 4; LRRC64; LRRIQ4
Gene Symbols LRRIQ4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRIQ4. Recombinant protein control fragment (Product #RP-108817).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 344657
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.