Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRK2 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11055289C
Description
LRRK2 Polyclonal specifically detects LRRK2 in Mouse, Human (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LRRK2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
augmented in rheumatoid arthritis 17, AURA17, Dardarin, DKFZp434H2111, EC 2.7.11.1, FLJ45829, leucine-rich repeat kinase 2, leucine-rich repeat serine/threonine-protein kinase 2, PARK8DARDARIN, Parkinson disease (autosomal dominant) 8, RIPK7, ROCO2 | |
Rabbit | |
285 kDa | |
0.1 mL | |
Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
120892 | |
Mouse, Human (Negative) | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LRRK2 | |
A synthetic peptide made to an internal region of the mouse LRRK2 protein sequence (between residues 900-1000). [Swiss-Prot: Q5S006] | |
Affinity Purified | |
RUO | |
Primary | |
Mouse. Does not react with Human. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction