Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRK2 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300268JF646
Description
LRRK2 Polyclonal specifically detects LRRK2 in Human, Mouse, Bovine, C. elegans, Insect, Plant samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.Specifications
LRRK2 | |
Polyclonal | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated | |
augmented in rheumatoid arthritis 17, AURA17, Dardarin, DKFZp434H2111, EC 2.7.11.1, FLJ45829, leucine-rich repeat kinase 2, leucine-rich repeat serine/threonine-protein kinase 2, PARK8DARDARIN, Parkinson disease (autosomal dominant) 8, RIPK7, ROCO2 | |
Rabbit | |
Affinity Purified | |
Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase | |
120892.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LRRK2 | |
A C-terminal synthetic peptide made to the human LRRK2 protein sequence (between residues 2500-2527). [UniProt No. Q5S007] | |
0.1 mL | |
Primary | |
Human, Mouse, Bovine, C. elegans, Insect, Plant | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction