Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRK2 Antibody (SR1452), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | LRRK2 |
---|---|
Clone | SR1452 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
LRRK2 Monoclonal antibody specifically detects LRRK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
LRRK2 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
augmented in rheumatoid arthritis 17, AURA17, Dardarin, DKFZp434H2111, EC 2.7.11.1, FLJ45829, leucine-rich repeat kinase 2, leucine-rich repeat serine/threonine-protein kinase 2, PARK8DARDARIN, Parkinson disease (autosomal dominant) 8, RIPK7, ROCO2 | |
A synthesized peptide derived from human LRRK2 (Uniprot #: Q5S007) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1452 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
120892 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title