Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human LRRK2 [D1994A], GST Tag Recombinant Protein

Catalog No. PV6051
Encompass_Preferred
Click to view available options
Quantity:
10 μg

Recombinant Protein

LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain. The protein is present largely in the cytoplasm but also associates with the mitochondrial outer membrane. Mutations in LRRK2 (R1441C Mutant) have been associated with Parkinson disease-8.
TRUSTED_SUSTAINABILITY

Specifications

Concentration See Label
For Use With (Application) Kinase Assay
Formulation proprietary buffer with no preservative
Molecular Weight (g/mol) 205 kDa
Name Human LRRK2 [D1994A], GST Tag
Quantity 10 μg
Common Name LRRK2 (D1994A Mutant)
KinaseFamily LRRK Kinase Family
Protein Length 970-2527; D1994A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Kinase Group Ser⁄Thr Kinases: TKL Group
Shipping Condition Dry Ice
Target Entry LRRK2 D1994A
Target Gene LRRK2
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Catalytic, Mutant, Inactive
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction, Neurobiology
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.