Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ LRRK2-IN-1
GREENER_CHOICESDP

Catalog No. 427350 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-735-0 50 mg
42-731-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-735-0 Supplier R&D Systems™ Supplier No. 4273/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective LRRK2 inhibitor

Potent and selective inhibitor of leucine-rich repeat kinase 2 (LRRK2). Inhibits both G2019S mutant and wild-type LRRK2 kinase activity (IC50 values are 6 and 13 nM respectively). Causes dephosphorylation, ubiquitination and degradation of LRRK2. Inhibits IFN-γ-induced monocyte maturation in vitro.

Chemical Identifiers

CAS 1234480-84-2
Molecular Formula C31H38N8O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 570.09
Purity >98%
Molecular Weight (g/mol) 570.698
Chemical Name or Material LRRK2-IN-1
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.