Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ LRRK2 Inhibitor III, HG-10-102-01

Catalog No. 43819510mg Shop All MilliporeSigma Products
Click to view available options
Quantity:
10 mg

A cell-permeable 2,4-diaminopyrimidinyl compound that acts as a potent, ATP-competitive LRRK2-selective inhibitor (IC5050 =

A cell-permeable 2,4-diaminopyrimidinyl compound that acts as a potent, ATP-competitive LRRK2-selective inhibitor (IC5050 = 20.3, 3.2, 153.7 and 95.9 nM, respectively,against human wt LRRK2 and G2019S, A2016T, G2019S/A2016T LRRK2 mutant; [ATP] = 100μM), displaying much reduced potency against MNK2 and MLK1 (IC50 = 0.6 and 2.1μM, respectively; [ATP] = 100μM) and little activity toward a panel of 136 other kinases. Although both HG-10-102-01 and LRRK2-In-1 438193) inhibit cellular wt and G20195 LRRK2 phosphorylation (Optimal conc. 1 to 3μM), only HG-10-102-01 is effective against cellular A2016T and G2019S/A2016T LRRK2 phosphorylations (IC50 <3μM). And only HG-10-102-01, but not LRRK2-In-1 or CZC-25146 (Cat. No. 438194), can cross blood-brain-barrier for LRRK2 phosphorylation inhibition in mice (30mg/kg to 50mg/kg i.p.) in vivo .
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 99% by HPLC
Target Proteins LRRK2
Molecular Formula C17H20ClN5O3
Formula Weight 377.8g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.