Learn More
Promega LRRK2 Kinase Enzyme System, 10microg
Catalog No. PRV4474 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4474 10 μg
1 options
Catalog No. PRV4474 Supplier Promega Supplier No. V4474
LRRK2 or leucine-rich repeat kinase is a protein with an ankryin repeat region, a leucine-rich repeat (LRR) domain, a kinase domain, a DFG-like motif, a RAS domain, a GTPase domain, a MLK-like domain and a WD40 domain.
  • Easily Screen and Profile LRRK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C