Learn More
Promega LRRK2 Kinase Enzyme System, 1mg
Catalog No. PRV6442 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6442 1 mg
1 options
Catalog No. PRV6442 Supplier Promega Supplier No. V6442
Add to Cart
Add to Cart
Request a Quote
LRRK2 or leucine-rich repeat kinase is a protein with an ankryin repeat region, a leucine-rich repeat (LRR) domain, a kinase domain, a DFG-like motif, a RAS domain, a GTPase domain, a MLK-like domain and a WD40 domain.
  • Easily Screen and Profile LRRK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C