Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRK2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585363
Description
Keep as concentrated solution. Predicted reactivity: Mouse (82%), Rat (80%), Pig (92%), Bovine (87%). Positive Control: NT2D1, IMR32, U87-MG, MCF-7, mouse cerebellum. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain. Mutations in LRRK2 have been associated with Parkinson's disease. LRRK2 is a member of the leucine-rich repeat kinase family and encodes a protein with an ankryin repeat region, a leucine-rich repeat domain, a kinase domain, a DFG-like motif, a RAS domain, a GTPase domain, a MLK-like domain, and a WD40 domain. The protein is present largely in the cytoplasm but also associates with the mitochondrial outer membrane.
Specifications
LRRK2 | |
Polyclonal | |
Unconjugated | |
LRRK2 | |
4921513O20Rik; 9330188B09Rik; augmented in rheumatoid arthritis 17; AURA17; AW561911; BC 300-268; cI-46; D630001M17Rik; DARDARIN; DKFZp434H2111; FLJ45829; Gm927; leucine rich repeat kinase 2; leucine-rich repeat kinase 2; leucine-rich repeat serine/threonine-protein kinase 2; LRR kinase 2; LRRK 2; Lrrk2; LRRK-2; PARK8; RIPK7; ROCO2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
120892, 66725 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q5S006, Q5S007 | |
LRRK2 | |
Recombinant protein encompassing a sequence within the center region of human LRRK2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction