Learn More
Invitrogen™ Human LRRK2, GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots; aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Related Links: LRRK2 tools for advancing Parkinson's disease research Kinase protein portfolio LanthaScreen™ Eu Kinase Binding Assay LanthaScreen™ Activity Assay Protein Form: Recombinant, Catalytic.
Specifications
Specifications
| Accession Number | Q5S007 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM tris with 2mM DTT, 50% glycerol, 150mM NaCl, 0.02% polysorbate 20, 0.5mM EDTA and no preservative; pH 7.5 |
| Gene ID (Entrez) | 120892 |
| Molecular Weight (g/mol) | 204.9 kDa |
| Name | Human LRRK2, GST Tag |
| pH Range | 7.5 |
| Purification Method | Purified |
| Quantity | 10 μg |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.