Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRK2 (WT) Recombinant Protein

Catalog No. p-7121791
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 μg
100 μg

Recombinant Protein

LRRK2 (Leucine Rich Repeat Kinase 2) is a Protein Coding gene. Diseases associated with LRRK2 include Parkinson Disease 8, Autosomal Dominant and Hereditary Late-Onset Parkinson Disease. Among its related pathways are Signaling by PTK6 and ncRNAs involved in Wnt signaling in hepatocellular carcinoma. Gene Ontology (GO) annotations related to LRRK2 (G2019S Mutant) include protein homodimerization activity and transferase activity, transferring phosphorus-containing groups. An important paralog of LRRK2 (G2019S Mutant) is TNNI3K. Mutations in the leucine-rich repeat kinase 2 (LRRK2) gene, which codes for a multidomain protein kinase, are considered the most common genetic cause of late-onset autosomal-dominant Parkinson's disease.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5S007
Concentration See Label
For Use With (Application) Kinase Assay
Formulation proprietary buffer with no preservative
Gene ID (Entrez) 120892
Molecular Weight (g/mol) 288 kDa
Name LRRK2 (WT)
Quantity 100 μg
Gene Alias LRRK2
Common Name LRRK2 (WT)
KinaseFamily LRRK Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag DYKDDDDK-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Research Category Neurobiology
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.