Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRN2 Monoclonal Antibody (6A9)
GREENER_CHOICE

Catalog No. PIMA521045
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521045 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521045 Supplier Invitrogen™ Supplier No. MA521045
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: RVQETHPYHI LLSWVTPPNT VSTNLTWSSA SSLRGQGATA LARLPRGTHS YNITRLLQAT EYWACLQVAF ADAHTQLACV WARTKEATSC HRALGDRPG.

The protein encoded by this gene belongs to the leucine-rich repeat superfamily. This gene was found to be amplified and overexpressed in malignant gliomas. The encoded protein has homology with other proteins that function as cell-adhesion molecules or as signal transduction receptors and is a candidate for the target gene in the 1q32.1 amplicon in malignant gliomas. Two alternatively spliced transcript variants encoding the same protein have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRN2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6A9
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene LRRN2
Gene Accession No. O75325
Gene Alias 5730406J09Rik; fibronectin type III, immunoglobulin and leucine rich repeat domain 7; FIGLER7; GAC1; Glioma amplified on chromosome 1 protein; leucine rich and ankyrin repeats 1; leucine rich repeat neuronal 2; leucine rich repeat neuronal 5; leucine rich repeat protein 2, neuronal; leucine-rich repeat neuronal protein 2; Leucine-rich repeat neuronal protein 5; LRANK1; LRRN2; LRRN5; NLRR-2; UNQ256/PRO293
Gene Symbols LRRN2
Host Species Mouse
Immunogen LRRN2 (NP_006329, 532 a.a. ~ 630 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10446
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.