Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRN2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA527923

Catalog No. PIPA527923


Only null left
Add to Cart

Description

Description

Recommended positive controls: U87-MG. Predicted reactivity: Mouse (93%), Rat (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene belongs to the leucine-rich repeat superfamily. This gene was found to be amplified and overexpressed in malignant gliomas. The encoded protein has homology with other proteins that function as cell-adhesion molecules or as signal transduction receptors and is a candidate for the target gene in the 1q32.1 amplicon in malignant gliomas. Two alternatively spliced transcript variants encoding the same protein have been described for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LRRN2
Polyclonal
Unconjugated
LRRN2
5730406J09Rik; fibronectin type III, immunoglobulin and leucine rich repeat domain 7; FIGLER7; GAC1; Glioma amplified on chromosome 1 protein; leucine rich and ankyrin repeats 1; leucine rich repeat neuronal 2; leucine rich repeat neuronal 5; leucine rich repeat protein 2, neuronal; leucine-rich repeat neuronal protein 2; Leucine-rich repeat neuronal protein 5; LRANK1; LRRN2; LRRN5; NLRR-2; UNQ256/PRO293
Rabbit
Antigen affinity chromatography
RUO
10446
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
O75325
LRRN2
Recombinant fragment corresponding to a region within amino acids 213 and 575 of Human LRRN2
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.