Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRN3 Monoclonal Antibody (A4-6-3)
GREENER_CHOICE

Catalog No. PIMA542566
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542566 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542566 Supplier Invitrogen™ Supplier No. MA542566
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Positive Control: SHSY5Ycell lysate, rat testis tissue lysate, F9. Subcellular Location: Membrane.

TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRN3
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone A4-6-3
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene LRRN3
Gene Accession No. Q8CBC6, Q9ESY6, Q9H3W5
Gene Alias fibronectin type III, immunoglobulin and leucine rich repeat domains 5; FIGLER5; leucine rich repeat neuronal 3; leucine rich repeat protein 3, neuronal; leucine-rich repeat neuronal protein 3; leucine-rich repeat protein, neuronal 3; Lrrn3; Nbla10363; Neuronal leucine-rich repeat protein 3; NLRR3; NLRR-3; UNQ194/PRO220
Gene Symbols LRRN3
Host Species Mouse
Immunogen Recombinant protein within human LRRN3 aa 1-260.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16981, 54674, 81514
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.