Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRN3/NLRR-3 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299841H
Description
LRRN3/NLRR-3 Polyclonal specifically detects LRRN3/NLRR-3 in Human samples. It is validated for ELISA.Specifications
LRRN3/NLRR-3 | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
54674 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
HRP | |
FIGLER5, immunoglobulin and leucine rich repeat domains 5, leucine rich repeat neuronal 3, leucine-rich repeat neuronal protein 3, leucine-rich repeat protein, neuronal 3, Neuronal leucine-rich repeat protein 3, NLRR3, NLRR-3 | |
Produced in rabbits immunized with purified, recombinant LRRN3/NLRR-3 (AAH35133.1; Met1-Thr628). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction