Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRN4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567401
Description
Immunogen sequence: SNSSVFPRAAS TTRTQHRGEH APELVLEPDI SAASTPLASK LLGPFPTSWD RSISSPQPGQ RTHATPQAPN PSLSEGEIPV LLLDDY Highest antigen sequence identity to the following orthologs - mouse 49%, rat 49%.
LRRN4 may play an important role in hippocampus-dependent long-lasting memory.
Specifications
LRRN4 | |
Polyclonal | |
Unconjugated | |
LRRN4 | |
C20orf75; dJ1056H1.1; leucine rich repeat neuronal 4; leucine-rich repeat neuronal protein 4; LRRN4; Neuronal leucine-rich repeat protein 4; NLRR4; NLRR-4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
164312 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WUT4 | |
LRRN4 | |
Recombinant Human LRRN4. Recombinant protein control fragment (Product #RP-108065). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction