Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRN4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571435
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571435 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571435 Supplier Invitrogen™ Supplier No. PA571435
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRRN4 Polyclonal Antibody for Western Blot

LRRN4 may play an important role in hippocampus-dependent long-lasting memory.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRN4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LRRN4
Gene Accession No. Q8WUT4
Gene Alias C20orf75; dJ1056H1.1; leucine rich repeat neuronal 4; leucine-rich repeat neuronal protein 4; LRRN4; Neuronal leucine-rich repeat protein 4; NLRR4; NLRR-4
Gene Symbols LRRN4
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human LRRN4.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 164312
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.