Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRN4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532907
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532907 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532907 Supplier Invitrogen™ Supplier No. PA532907
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan (100%); Marmoset (94%). Immunohistochemistry with formalin-fixed paraffin-embedded tissues requires pretreatment using HIER with citrate buffer.

LRRN4 may play an important role in hippocampus-dependent long-lasting memory.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRN4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene LRRN4
Gene Accession No. Q8WUT4
Gene Alias C20orf75; dJ1056H1.1; leucine rich repeat neuronal 4; leucine-rich repeat neuronal protein 4; LRRN4; Neuronal leucine-rich repeat protein 4; NLRR4; NLRR-4
Gene Symbols LRRN4
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from N-Terminus of human LRRN4 / C20orf75.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 164312
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.