Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRN4CL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562168
Description
Immunogen sequence: QDFEEEEADE TETAWPPLPA VPCDYDHCRH LQVPCKELQR VGPAACLCPG LSSPAQPPDP PRMGEVRIAA EEGRAVVHWC A Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 63%.

Specifications
LRRN4CL | |
Polyclonal | |
Unconjugated | |
LRRN4CL | |
LRRN4 C-terminal like; LRRN4 C-terminal-like protein; LRRN4CL; UNQ728/PRO1410 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
221091 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.15 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8ND94 | |
LRRN4CL | |
Recombinant protein corresponding to Human LRRN4CL. Recombinant protein control fragment (Product #RP-99930). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction