Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRTM1 Antibody (OTI5H9), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272543H
Description
LRRTM1 Monoclonal specifically detects LRRTM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
LRRTM1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
LRRTM1 | |
Recombinant protein fragment corresponding to amino acids 182-422 of human LRRTM1 (NP_849161) produced in E.coli. | |
0.1 mL | |
347730.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI5H9 | |
Western Blot, Immunohistochemistry | |
FLJ32082, leucine rich repeat transmembrane neuronal 1, leucine-rich repeat transmembrane neuronal 1 protein, leucine-rich repeat transmembrane neuronal protein 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction