Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRTM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119942
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119942 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119942 Supplier Invitrogen™ Supplier No. PA5119942
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: SH-SY5Y cell lysate, N2A cell lysate, rat brain tissue lysate, N2A, SH-SY5Y. Subcellular Location: Cell membrane, postsynaptic cell membrane.

The Leucine-rich repeat transmembrane neuronal proteins (LRRTMs) are differentially expressed in the nervous system and were recently found to instruct presynaptic and mediate postsynaptic glutamatergic differentiation, with LRRTM1 and LRRTM2 most potent at inducing presynaptic differentiation. Each LRRTM protein is a type I transmembrane containing ten extracellular leucine-rich repeats and a short intracellular tail and has a developmentally regulated pattern distinct from all others. LRRTM1 is a maternally suppressed gene that is associated paternally with handedness and schizophrenia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRTM1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 50% glycerol and 0.05% sodium azide; pH 7.4
Gene LRRTM1
Gene Accession No. D4A6D8, Q86UE6, Q8K377
Gene Alias 4632401D06Rik; AW125451; leucine rich repeat transmembrane neuronal 1; leucine-rich repeat transmembrane neuronal 1; leucine-rich repeat transmembrane neuronal 1 protein; leucine-rich repeat transmembrane neuronal protein 1; Lrrtm1; UNQ675/PRO1309
Gene Symbols LRRTM1
Host Species Rabbit
Immunogen Synthetic peptide within mouse LRRTM1 aa 320-360 (Extracellular).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347730, 679668, 74342
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.